Anti-RPS6KB1 Mouse Monoclonal Antibody [clone: 215247]

Supplier: R & D Systems
MAB8962
220021-637EA 655.11 USD
220021-637
Anti-RPS6KB1 Mouse Monoclonal Antibody [clone: 215247]
Antibodies
Type: Primary
Antigen: RPS6KB1
Clonality: Monoclonal
Clone: 215247
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR