Anti-MERTK Mouse Monoclonal Antibody [clone: 125508]

Supplier: R & D Systems
MAB8911
220021-635EA 763.57 USD
220021-635
Anti-MERTK Mouse Monoclonal Antibody [clone: 125508]
Antibodies
Type: Primary
Antigen: MERTK
Clonality: Monoclonal
Clone: 125508
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR