Anti-MERTK Mouse Monoclonal Antibody [clone: 125520]

Supplier: R & D Systems
MAB891
220021-634EA 772.25 USD
220021-634
Anti-MERTK Mouse Monoclonal Antibody [clone: 125520]
Antibodies
Type: Primary
Antigen: MERTK
Clonality: Monoclonal
Clone: 125520
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR