Anti-APP Mouse Monoclonal Antibody [clone: 128715]

Supplier: R & D Systems
MAB890
220021-633EA 763.57 USD
220021-633
Anti-APP Mouse Monoclonal Antibody [clone: 128715]
Antibodies
Type: Primary
Antigen: APP
Clonality: Monoclonal
Clone: 128715
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR