Anti-AKT (pan specific) Mouse Monoclonal Antibody [clone: 5457]

Supplier: R & D Systems
MAB887
102887-624EA 616.06 USD
102887-624
Anti-AKT (pan specific) Mouse Monoclonal Antibody [clone: 5457]
Antibodies
Simple Western

Type: Primary
Antigen: AKT (pan specific)
Clonality: Monoclonal
Clone: 5457
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR