Anti-BIRC5 Mouse Monoclonal Antibody [clone: 91630]

Supplier: R & D Systems
MAB886
220021-632EA 594.37 USD
220021-632
Anti-BIRC5 Mouse Monoclonal Antibody [clone: 91630]
Antibodies
Type: Primary
Antigen: BIRC5
Clonality: Monoclonal
Clone: 91630
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR