Anti-TNFSF13 Mouse Monoclonal Antibody [clone: 670840]

Supplier: R & D Systems
MAB8843
102887-622EA 642.09 USD
102887-622
Anti-TNFSF13 Mouse Monoclonal Antibody [clone: 670840]
Antibodies
Type: Primary
Antigen: TNFSF13
Clonality: Monoclonal
Clone: 670840
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR