Anti-BCL2L4 Mouse Monoclonal Antibody [clone: 127602]

Supplier: R & D Systems
MAB883
220021-629EA 561.11 USD
220021-629
Anti-BCL2L4 Mouse Monoclonal Antibody [clone: 127602]
Antibodies
Type: Primary
Antigen: BCL2L4
Clonality: Monoclonal
Clone: 127602
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR