Anti-DYNLL1 Mouse Monoclonal Antibody [clone: 98618]

Supplier: R & D Systems
MAB877
220021-627EA 763.57 USD
220021-627
Anti-DYNLL1 Mouse Monoclonal Antibody [clone: 98618]
Antibodies
Type: Primary
Antigen: DYNLL1
Clonality: Monoclonal
Clone: 98618
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR