Anti-IGFBP6 Mouse Monoclonal Antibody [clone: 110211]

Supplier: R & D Systems
MAB8761
220021-625EA 772.25 USD
220021-625
Anti-IGFBP6 Mouse Monoclonal Antibody [clone: 110211]
Antibodies
Type: Primary
Antigen: IGFBP6
Clonality: Monoclonal
Clone: 110211
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR