Anti-IL10RB Mouse Monoclonal Antibody [clone: 90227]

Supplier: R & D Systems
MAB8741
220021-622EA 772.25 USD
220021-622
Anti-IL10RB Mouse Monoclonal Antibody [clone: 90227]
Antibodies
Type: Primary
Antigen: IL10RB
Clonality: Monoclonal
Clone: 90227
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR