Anti-LSAMP Mouse Monoclonal Antibody [clone: 110722]

Supplier: R & D Systems
MAB8731
220021-620EA 763.57 USD
220021-620
Anti-LSAMP Mouse Monoclonal Antibody [clone: 110722]
Antibodies
Type: Primary
Antigen: LSAMP
Clonality: Monoclonal
Clone: 110722
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR