Anti-MAPK14 Mouse Monoclonal Antibody [clone: 696602]

Supplier: R & D Systems
MAB8691
102887-620EA 594.37 USD
102887-620
Anti-MAPK14 Mouse Monoclonal Antibody [clone: 696602]
Antibodies
Type: Primary
Antigen: MAPK14
Clonality: Monoclonal
Clone: 696602
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR