Anti-MAPK14 Mouse Monoclonal Antibody [clone: 142102]

Supplier: R & D Systems
MAB869
220021-617EA 594.37 USD
220021-617
Anti-MAPK14 Mouse Monoclonal Antibody [clone: 142102]
Antibodies
Type: Primary
Antigen: MAPK14
Clonality: Monoclonal
Clone: 142102
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR