Anti-APAF1 Mouse Monoclonal Antibody [clone: 9448]

Supplier: R & D Systems
MAB868
220021-616EA 577.02 USD
220021-616
Anti-APAF1 Mouse Monoclonal Antibody [clone: 9448]
Antibodies
Type: Primary
Antigen: APAF1
Clonality: Monoclonal
Clone: 9448
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR