Anti-CXCL14 Mouse Monoclonal Antibody [clone: 131120]

Supplier: R & D Systems
MAB866
220021-614EA 779.48 USD
220021-614
Anti-CXCL14 Mouse Monoclonal Antibody [clone: 131120]
Antibodies
Type: Primary
Antigen: CXCL14
Clonality: Monoclonal
Clone: 131120
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR