Anti-PDPK1 Mouse Monoclonal Antibody [clone: 650308]

Supplier: R & D Systems
MAB864
102887-618EA 594.37 USD
102887-618
Anti-PDPK1 Mouse Monoclonal Antibody [clone: 650308]
Antibodies
Type: Primary
Antigen: PDPK1
Clonality: Monoclonal
Clone: 650308
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR