Anti-BID Rat Monoclonal Antibody [clone: 91508]

Supplier: R & D Systems
MAB860
220021-612EA 633.42 USD
220021-612
Anti-BID Rat Monoclonal Antibody [clone: 91508]
Antibodies
Type: Primary
Antigen: BID
Clonality: Monoclonal
Clone: 91508
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG1
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR