Anti-C19ORF80 Mouse Monoclonal Antibody [clone: 910248]

Supplier: R & D Systems
MAB8548
MSPP-MAB8548EA 647.88 USD
MSPP-MAB8548
Anti-C19ORF80 Mouse Monoclonal Antibody [clone: 910248]
Antibodies
Type: Primary
Antigen: C19ORF80
Clonality: Monoclonal
Clone: 910248
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR