Anti-AXL Rat Monoclonal Antibody [clone: 175128]

Supplier: R & D Systems
MAB8541
220021-609EA 639.2 USD
220021-609
Anti-AXL Rat Monoclonal Antibody [clone: 175128]
Antibodies
Type: Primary
Antigen: AXL
Clonality: Monoclonal
Clone: 175128
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR