Anti-AXL Rat Monoclonal Antibody [clone: 107332]

Supplier: R & D Systems
MAB854
220021-608EA 763.57 USD
220021-608
Anti-AXL Rat Monoclonal Antibody [clone: 107332]
Antibodies
Type: Primary
Antigen: AXL
Clonality: Monoclonal
Clone: 107332
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG1
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR