Anti-GDF5 Rat Monoclonal Antibody [clone: 143004]

Supplier: R & D Systems
MAB8531
220021-607EA 772.25 USD
220021-607
Anti-GDF5 Rat Monoclonal Antibody [clone: 143004]
Antibodies
Type: Primary
Antigen Symbol: GDF5
Antigen Name: growth differentiation factor 5
Antigen Synonyms: BMP-14, Cdmp-1, bp, brp
Clonality: Monoclonal
Clone: 143004
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR