Anti-TNFRSF8 Rat Monoclonal Antibody [clone: 115705]

Supplier: R & D Systems
MAB8521
220021-605EA 772.25 USD
220021-605
Anti-TNFRSF8 Rat Monoclonal Antibody [clone: 115705]
Antibodies
Type: Primary
Antigen: TNFRSF8
Clonality: Monoclonal
Clone: 115705
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR