Anti-BAG1 Mouse Monoclonal Antibody [clone: GP3.10G3E2]

Supplier: R & D Systems
MAB852
220021-604EA 561.11 USD
220021-604
Anti-BAG1 Mouse Monoclonal Antibody [clone: GP3.10G3E2]
Antibodies
Type: Primary
Antigen: BAG1
Clonality: Monoclonal
Clone: GP3.10G3E2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR