Anti-BAD Rat Monoclonal Antibody [clone: 64103]

Supplier: R & D Systems
MAB850
220021-602EA 566.89 USD
220021-602
Anti-BAD Rat Monoclonal Antibody [clone: 64103]
Antibodies
Type: Primary
Antigen: BAD
Clonality: Monoclonal
Clone: 64103
Conjugation: Unconjugated
Epitope: N-Terminal
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR