Anti-PTEN Mouse Monoclonal Antibody [clone: 217702]

Supplier: R & D Systems
MAB847
220021-600EA 639.2 USD
220021-600
Anti-PTEN Mouse Monoclonal Antibody [clone: 217702]
Antibodies
Type: Primary
Antigen: PTEN
Clonality: Monoclonal
Clone: 217702
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR