Anti-BCL2L4 Mouse Monoclonal Antibody [clone: 127606]

Supplier: R & D Systems
MAB846
220021-599EA 577.02 USD
220021-599
Anti-BCL2L4 Mouse Monoclonal Antibody [clone: 127606]
Antibodies
Type: Primary
Antigen: BCL2L4
Clonality: Monoclonal
Clone: 127606
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR