Anti-CCL1 Rat Monoclonal Antibody [clone: 148107]

Supplier: R & D Systems
MAB8451
220021-598EA 772.25 USD
220021-598
Anti-CCL1 Rat Monoclonal Antibody [clone: 148107]
Antibodies
Type: Primary
Antigen: CCL1
Clonality: Monoclonal
Clone: 148107
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype:
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR