Anti-IL18R1 Mouse Monoclonal Antibody [clone: 70614]

Supplier: R & D Systems
MAB8401
220021-596EA 772.25 USD
220021-596
Anti-IL18R1 Mouse Monoclonal Antibody [clone: 70614]
Antibodies
Type: Primary
Antigen: IL18R1
Clonality: Monoclonal
Clone: 70614
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR