Anti-HHEX Mouse Monoclonal Antibody [clone: 906705]

Supplier: R & D Systems
MAB8377
102887-616EA 577.02 USD
102887-616
Anti-HHEX Mouse Monoclonal Antibody [clone: 906705]
Antibodies
Simple Western

Type: Primary
Antigen: HHEX
Clonality: Monoclonal
Clone: 906705
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR