Anti-SLC12A5 Mouse Monoclonal Antibody [clone: 905705]

Supplier: R & D Systems
MAB8369
102887-614EA 600.15 USD
102887-614
Anti-SLC12A5 Mouse Monoclonal Antibody [clone: 905705]
Antibodies
Type: Primary
Antigen: SLC12A5
Clonality: Monoclonal
Clone: 905705
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR