Anti-CASP3 Rabbit Monoclonal Antibody [clone: 269518]

Supplier: R & D Systems
MAB835
220021-592EA 582.8 USD
220021-592
Anti-CASP3 Rabbit Monoclonal Antibody [clone: 269518]
Antibodies
Type: Primary
Antigen: CASP3
Clonality: Monoclonal
Clone: 269518
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR