Anti-IMPDH2 Mouse Monoclonal Antibody [clone: 911425]

Supplier: R & D Systems
MAB8349
102887-610EA 594.37 USD
102887-610
Anti-IMPDH2 Mouse Monoclonal Antibody [clone: 911425]
Antibodies
Simple Western

Type: Primary
Antigen: IMPDH2
Clonality: Monoclonal
Clone: 911425
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR