Anti-CASP10 Mouse Monoclonal Antibody [clone: 63131]

Supplier: R & D Systems
MAB834
220021-591EA 577.02 USD
220021-591
Anti-CASP10 Mouse Monoclonal Antibody [clone: 63131]
Antibodies
Type: Primary
Antigen: CASP10
Clonality: Monoclonal
Clone: 63131
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR