Anti-EGFR Mouse Monoclonal Antibody [clone: 752502]

Supplier: R & D Systems
MAB8336
102887-608EA 594.37 USD
102887-608
Anti-EGFR Mouse Monoclonal Antibody [clone: 752502]
Antibodies
Simple Western

Type: Primary
Antigen: EGFR
Clonality: Monoclonal
Clone: 752502
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR