Anti-SLC18A2 Mouse Monoclonal Antibody [clone: 899327]

Supplier: R & D Systems
MAB8327
102887-606EA 594.37 USD
102887-606
Anti-SLC18A2 Mouse Monoclonal Antibody [clone: 899327]
Antibodies
Type: Primary
Antigen: SLC18A2
Clonality: Monoclonal
Clone: 899327
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR