Anti-B2M Rat Monoclonal Antibody [clone: 893803]

Supplier: R & D Systems
MAB8325
102887-604EA 633.42 USD
102887-604
Anti-B2M Rat Monoclonal Antibody [clone: 893803]
Antibodies
Type: Primary
Antigen: B2M
Clonality: Monoclonal
Clone: 893803
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR