Anti-CCR6 Mouse Monoclonal Antibody [clone: 876515]

Supplier: R & D Systems
MAB8320
102887-602EA 561.11 USD
102887-602
Anti-CCR6 Mouse Monoclonal Antibody [clone: 876515]
Antibodies
Type: Primary
Antigen: CCR6
Clonality: Monoclonal
Clone: 876515
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR