Anti-H2AF12M Mouse Monoclonal Antibody [clone: 866416]

Supplier: R & D Systems
MAB8318
102887-600EA 577.02 USD
102887-600
Anti-H2AF12M Mouse Monoclonal Antibody [clone: 866416]
Antibodies
Type: Primary
Antigen Symbol: H2AF12M
Antigen Name: Histone H2AY
Antigen Synonyms: H2AFJ, H2AFY, Histone MacroH2A1, Medulloblastoma Antigen MU-MB-50.205
Clonality: Monoclonal
Clone: 866416
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR