Anti-L1TD1 Mouse Monoclonal Antibody [clone: 901258]

Supplier: R & D Systems
MAB8317
102887-598EA 600.15 USD
102887-598
Anti-L1TD1 Mouse Monoclonal Antibody [clone: 901258]
Antibodies
Type: Primary
Antigen: L1TD1
Clonality: Monoclonal
Clone: 901258
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR