Anti-AGRP Mouse Monoclonal Antibody [clone: 8286]

Supplier: R & D Systems
MAB8311
220021-589EA 763.57 USD
220021-589
Anti-AGRP Mouse Monoclonal Antibody [clone: 8286]
Antibodies
Type: Primary
Antigen: AGRP
Clonality: Monoclonal
Clone: 8286
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR