Anti-RAB13 Mouse Monoclonal Antibody [clone: 863028]

Supplier: R & D Systems
MAB8305
102887-596EA 577.02 USD
102887-596
Anti-RAB13 Mouse Monoclonal Antibody [clone: 863028]
Antibodies
Type: Primary
Antigen: RAB13
Clonality: Monoclonal
Clone: 863028
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR