Anti-CASP9 Mouse Monoclonal Antibody [clone: LAP6]

Supplier: R & D Systems
MAB8301
220021-588EA 594.37 USD
220021-588
Anti-CASP9 Mouse Monoclonal Antibody [clone: LAP6]
Antibodies
Simple Western

Type: Primary
Antigen: CASP9
Clonality: Monoclonal
Clone: LAP6
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR