Anti-NAIP Mouse Monoclonal Antibody [clone: 541609]

Supplier: R & D Systems
MAB829
102887-240EA 594.37 USD
102887-240
Anti-NAIP Mouse Monoclonal Antibody [clone: 541609]
Antibodies
Type: Primary
Antigen: NAIP
Clonality: Monoclonal
Clone: 541609
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR