Anti-CCR9 Mouse Monoclonal Antibody [clone: 882416]

Supplier: R & D Systems
MAB8287
102887-238EA 577.02 USD
102887-238
Anti-CCR9 Mouse Monoclonal Antibody [clone: 882416]
Antibodies
Type: Primary
Antigen: CCR9
Clonality: Monoclonal
Clone: 882416
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR