Anti-DRD5 Mouse Monoclonal Antibody [clone: 889040]

Supplier: R & D Systems
MAB8286
102887-236EA 561.11 USD
102887-236
Anti-DRD5 Mouse Monoclonal Antibody [clone: 889040]
Antibodies
Type: Primary
Antigen: DRD5
Clonality: Monoclonal
Clone: 889040
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR