Anti-MCL1 Mouse Monoclonal Antibody [clone: 602901]

Supplier: R & D Systems
MAB8281
102887-234EA 577.02 USD
102887-234
Anti-MCL1 Mouse Monoclonal Antibody [clone: 602901]
Antibodies
Type: Primary
Antigen: MCL1
Clonality: Monoclonal
Clone: 602901
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR