Anti-MCL1 Mouse Monoclonal Antibody [clone: 542808]

Supplier: R & D Systems
MAB828
220021-587EA 633.42 USD
220021-587
Anti-MCL1 Mouse Monoclonal Antibody [clone: 542808]
Antibodies
Type: Primary
Antigen: MCL1
Clonality: Monoclonal
Clone: 542808
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR