Anti-ACTN1 Mouse Monoclonal Antibody [clone: 9131]

Supplier: R & D Systems
MAB8279
102887-592EA 594.37 USD
102887-592
Anti-ACTN1 Mouse Monoclonal Antibody [clone: 9131]
Antibodies
Simple Western

Type: Primary
Antigen: ACTN1
Clonality: Monoclonal
Clone: 9131
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR