Anti-SMTN (B) Mouse Monoclonal Antibody [clone: 885424]

Supplier: R & D Systems
MAB8278
102887-558EA 577.02 USD
102887-558
Anti-SMTN (B) Mouse Monoclonal Antibody [clone: 885424]
Antibodies
Type: Primary
Antigen: SMTN (B)
Clonality: Monoclonal
Clone: 885424
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR